Merck
  • Asp3Gly polymorphism affects fatty acid-binding protein 3 intracellular stability and subcellular localization.
Asp3Gly polymorphism affects fatty acid-binding protein 3 intracellular stability and subcellular localization.
FEBS letters (2015-07-25)
Tatsuya Kusudo, Yasuhiko Hashida, Fujiko Ando, Hiroshi Shimokata, Hitoshi Yamashita
ABSTRACT
Fatty acid-binding proteins (FABP) play a crucial role in intracellular fatty acid transportation and metabolism. In this study, we investigate the effects of the FABP3 Asp3Gly (D3G) polymorphism on protein structure and function. Although the mutation did not alter protein secondary structure or the ability to bind 1-anilinonaphthalene-8-sulfonic acid and palmitate, the intracellular stability of the D3G mutant was significantly decreased. Immunocytochemical analysis reveals that the mutation alters FABP3 subcellular localization. Our results suggest that the D3G polymorphism may impact energy metabolism and physiological functions.
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
8-Anilino-1-naphthalenesulfonic acid
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Palmitic acid, natural, 98%, FG