Merck
  • Long-term imaging of Caenorhabditis elegans using nanoparticle-mediated immobilization.
Long-term imaging of Caenorhabditis elegans using nanoparticle-mediated immobilization.
PloS one (2013-01-10)
Eric Kim, Lin Sun, Christopher V Gabel, Christopher Fang-Yen
ABSTRACT
One advantage of the nematode Caenorhabditis elegans as a model organism is its suitability for in vivo optical microscopy. Imaging C. elegans often requires animals to be immobilized to avoid movement-related artifacts. Immobilization has been performed by application of anesthetics or by introducing physical constraints using glue or specialized microfluidic devices. Here we present a method for immobilizing C. elegans using polystyrene nanoparticles and agarose pads. Our technique is technically simple, does not expose the worm to toxic substances, and allows recovery of animals. We evaluate the method and show that the polystyrene beads increase friction between the worm and agarose pad. We use our method to quantify calcium transients and long-term regrowth in single neurons following axotomy by a femtosecond laser.
Sigma-Aldrich
2-Phenylethyl β-D-thiogalactoside, ≥98% (TLC)
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, for molecular biology
Sigma-Aldrich
Agarose, High EEO, for molecular biology