Skip to Content
IN
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Safety Data Sheets (SDS)
Certificates (COA/COO)
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
Transferpette
®
pipette, digital, adjustable single channel
Products
Product Category
pipettors (10)
Manufacturer
Volume (µL)
Electronic
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z328006
BRAND
®
Transferpette
®
pipette, digital, adjustable single channel,
volume 10-100 μL, accuracy: 0.6%, precision: 0.2%
Expand
View Pricing
Z328030
BRAND
®
Transferpette
®
pipette, digital, adjustable single channel,
volume 100-1000 μL, accuracy: 0.6%, precision: 0.2%
Expand
View Pricing
Z328022
BRAND
®
Transferpette
®
pipette, digital, adjustable single channel,
volume 25-250 μL, accuracy: 0.6%, precision: 0.2%
Expand
View Pricing
Z327999
BRAND
®
Transferpette
®
pipette, digital, adjustable single channel,
volume 5-50 μL, accuracy: 0.8%, precision: 0.4%
Expand
View Pricing
Z327964
BRAND
®
Transferpette
®
pipette, digital, adjustable single channel,
volume 0.5-10 μL, accuracy: 1.0%, precision: 0.8%
Expand
View Pricing
Z328049
BRAND
®
Transferpette
®
pipette, digital, adjustable single channel,
volume 500-5000 μL, accuracy: 0.6%, precision: 0.2%
Expand
View Pricing
Z328014
BRAND
®
Transferpette
®
pipette, digital, adjustable single channel,
volume 20-200 μL, accuracy: 0.6%, precision: 0.2%
Expand
View Pricing
Z327972
BRAND
®
Transferpette
®
pipette, digital, adjustable single channel,
volume 2-20 μL, accuracy: 0.8%, precision: 0.4%
Expand
View Pricing
Z327980
BRAND
®
Transferpette
®
pipette, digital, adjustable single channel,
volume 2-20 μL, accuracy: 0.8%, precision: 0.4%
Expand
View Pricing
Z327956
BRAND
®
Transferpette
®
pipette, digital, adjustable single channel,
volume 0.1-1 μL, accuracy: 2.0%, precision: 1.2%
Expand
View Pricing