
Bis(3,5-dimethylphenyl)phosphine

Empirical Formula (Hill Notation):
C16H19P
CAS Number:
Molecular Weight:
242.30
Products
Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing