Skip to Content
Merck
All Photos(3)

Key Documents

V900924

Sigma-Aldrich

D-Glucose 6-phosphate disodium salt hydrate

Vetec, reagent grade, ≥98%

Synonym(s):

D-Glucose 6-phosphate disodium salt hydrate, G-6-P-Na2, Robison ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11Na2O9P · xH2O
CAS Number:
Molecular Weight:
304.10 (anhydrous basis)
Beilstein:
5199009
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

≥98%

form

powder

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C6H13O9P.2Na.H2O/c7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;;;/h2-10H,1H2,(H2,11,12,13);;;1H2/q;2*+1;/p-2/t2-,3-,4+,5-,6?;;;/m1.../s1

InChI key

UUWJZXLTPORJKW-WYFATIGWSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service