Skip to Content
Merck
All Photos(1)

Key Documents

1A01530

USP

Aminophenylmorpholinone

Pharmaceutical Analytical Impurity (PAI)

Synonym(s):

(4-(4-aminophenyl)morpholin-3-one)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
₹1,14,701.70

₹1,14,701.70


Estimated to ship on05 June 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
₹1,14,701.70

About This Item

Empirical Formula (Hill Notation):
C10H12N2O2
CAS Number:
Molecular Weight:
192.21
MDL number:
UNSPSC Code:
41116100
NACRES:
NA.24

₹1,14,701.70


Estimated to ship on05 June 2025Details


Request a Bulk Order

grade

pharmaceutical analytical impurity (PAI)

Agency

USP

API family

rivaroxaban

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

SMILES string

N2(CCOCC2=O)c1ccc(cc1)N

InChI

1S/C10H12N2O2/c11-8-1-3-9(4-2-8)12-5-6-14-7-10(12)13/h1-4H,5-7,11H2

InChI key

MHCRLDZZHOVFEE-UHFFFAOYSA-N

General description

Aminophenylmorpholinone is a USP Pharmaceutical Analytical Impurity (PAI).

USP PAI are a product line of impurities suitable for research and analytical purposes, which help to ensure the quality and safety of medicines.

Associated Drug Substance: Rivaroxaban

Therapeutic Area: Anticoagulants & Antiplatelets.

For more information about this PAI, visit here.

Application

Aminophenylmorpholinone (USP PAI) is intended for use in analytical testing to detect, identify, and measure pharmaceutical impurities.

Features and Benefits

USP PAI advance your early analytical R&D and process development. PAI can be used in the following applications:
1. Conduct analytical tests during early formulation feasibility studies.
2. Determine degradation impurities produced during stress studies.
3. Develop, validate, and transfer analytical methods.
4. Perform spiking studies during process R&D to demonstrate depletion upon recrystallization.
5. Record retention times and/or spectra
6. Determine relative response factors.
7. Identify unknown impurities that formed during ICH stability conditions.
8. Identify impurities that are present in the Reference Listed Drug
9. Test for and profile impurities not listed in drug substance and drug product monographs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Order from a broad range of highly characterized primary reference standard materials to use with USP-NF monographs for the testing of drug substances & dosage forms, pharmaceutical excipients, food ingredients and dietary supplements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service