Skip to Content
Merck
All Photos(1)

Documents

N11502

Copper oxychloride

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116107

grade

technical grade

packaging

ampule of 1 g

manufacturer/tradename

Chem Service, Inc. PS-292

technique(s)

AAS: suitable
ICP: suitable

format

neat

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yvonne Willi et al.
Genetica, 139(7), 903-908 (2011-07-14)
The ability to improve fitness via adaptive evolution may be affected by environmental change. We tested this hypothesis in an in vitro experiment with the plant pathogen Rhizoctonia solani Anastomosis Group 3 (AG-3), assessing genetic and environmental variances under two
J Masaka et al.
The Science of the total environment, 387(1-3), 228-236 (2007-09-08)
A study was carried out at a sanitary waste disposal site for Kutsaga Tobacco Research Station, Zimbabwe, which uses large amounts of copper oxy chloride for sterilization of recycled float trays in flooded bench tobacco seedling production systems. Soil samples
E Pose-Juan et al.
Journal of hazardous materials, 174(1-3), 181-187 (2009-10-03)
The objective of this work is to asses the sorption of metalaxyl applied as a copper oxychloride (CO)-metalaxyl formulation, for a set of selected soils devoted to vineyards. The method involved batch incubation of soils suspended with a commercial copper
Olugbenga J Owojori et al.
Ecotoxicology and environmental safety, 73(3), 407-414 (2009-12-08)
We investigated the effect of clay content on the bioavailability of copper to the earthworm Eisenia fetida, in the laboratory for 28 days using OECD artificial soil adjusted to 5%, 20% and 40% clay. Mortality, growth, cocoon production and internal
Eva Pose et al.
Journal of agricultural and food chemistry, 57(7), 2843-2848 (2009-04-02)
The objective of this work was to assess the sorption of copper (Cu) applied as a Cu-oxychloride metalaxyl formulation by soils characterized by anthropogenic accumulation of Cu due to agricultural activity. The methods involved batch incubation of soils with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service