Skip to Content
Merck
All Photos(1)

Key Documents

G005796

LC/MS (TOF) Analysis of Illicit Bath Salts in Urine on Ascentis® Express HILIC after Solid Phase Extraction (SPE) using Supel-Select SCX

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

technique(s)

HPLC: suitable

test parameters

sample/matrix: urine samples spiked at 100 ng/mL with each target analyte (To ensure full ionization of the analytes, spiked samples were treated with formic acid to a final concentration of 0.1% formic acid.)
SPE tube/cartridge: Supel-Select SCX, 30 mg/1 mL (54240-U)
condition: 1 mL 1% formic acid acetonitrile then 1 mL water
sample addition: 1 mL spiked water blank or urine
washing: 1 mL water, 1 mL 1% formic acid acetonitrile, 1 mL water
elution: 2 mL 10% ammonium hydroxide in acetonitrile
eluate post-treatment: thoroughly mix via vortex agitation, evaporate 1 mL aliquot to dryness, reconstitute in 100 μL water:methanol
column: Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase: (A) 5 mM ammonium formate acetonitrile; (B) 5 mM ammonium formate water; (98:2, A:B); premixed
flow rate: 0.6 mL/min
pressure: 1842 psi (127 bar)
column temp.: 35 °C
detector: MS, ESI(+), TIC, m/z 100-1000
injection: 1 μL
sample: 200 μg/L ea. in acetonitrile

suitability

application for HPLC

application(s)

forensics and toxicology

Analysis Note

Bath salts are psychoactive designer drugs of the phenethylamine and cathinone families. Shown here is the rapid, sensitive LC/TOF-MS analysis of nine bath salts extracted from human urine using polymeric SPE on Supel-Select SCX and separated on an Ascentis Express HILIC column. Notice the lack of interfering peaks in the chromatogram demonstrating the effectiveness of the sample cleanup. Cerilliant CRMs were used to ensure reliable MS identification and quantification.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Supel is a trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    3,4-Methylenedioxypyrovalerone HCl (MDPV) solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    Buphedrone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    3-Fluoromethcathinone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    Butylone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    Ethylone hydrochloride, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    4-Fluoromethcathinone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    Mephedrone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    Methylone hydrochloride, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    Methedrone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    See All (9)

SPE tube or plate

mobile phase component


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service