Skip to Content
Merck
All Photos(1)

Documents

G005668

LC/MS Analysis of Antipsychotic Drugs on Ascentis® Express RP Amide

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express RP Amide, 10 cm x 2.1 mm I.D., 2.7 μm particles (53913-U)
mobile phase: [A] 5 mM ammonium formate water; [B] 5mM ammonium formate in 95:5 (v/v) methanol:water
gradient: held at 35% B for 0.5 min; 35 to 85% B in 5.5 min; held at 85% B for 1 min
flow rate: 0.3 mL/min
pressure: 4641 psi (320 bar)
column temp.: 35 °C
detector: ESI(+), m/z 100-1000
injection: 1 μL
sample: 200 ng/mL in waler

suitability

application for HPLC

application(s)

clinical
pharmaceutical (small molecule)

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    (2S,3S)-Hydroxybupropion hydrochloride, ≥98% (HPLC)
    Bupropion hydrochloride, United States Pharmacopeia (USP) Reference Standard
    Fluphenazine dihydrochloride

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service