Skip to Content
Merck
All Photos(1)

Key Documents

G005613

HPLC Analysis of Withania Extract on Ascentis® Express F5

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

Pricing and availability is not currently available.

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 μm particles (53569-U)
mobile phase: [A] water; [B] acetonitrile
gradient: 20 to 100% B in 5 min; held at 100% B for 0.5 min
flow rate: 0.3 mL/min
pressure: 2750 psi (190 bar)
column temp.: 35 °C
detector: UV, 227 nm
injection: 5 μL
sample: 20 mg/mL in water:acetonitrile (50:50); sonicate for 15 minutes; filter 0.45 μm; dilute to water:acetonitrile (80:20)

suitability

application for HPLC

application(s)

vitamins, nutraceuticals, and natural products

Analysis Note

This application demonstrates the suitability of Ascentis Express F5 for the efficient separation of withania extract.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Withanoside IV
    Withanoside V
    Withaferin A
    12-Deoxywithastramonolide
    Withanolide A
    Withanone
    Withanolide B

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service