Skip to Content
Merck
All Photos(1)

Key Documents

EPA 8270 Benzidines Mix

2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

CofA

certificate of analysis is enclosed in each package.

feature

standard type calibration

packaging

ampule of 1 mL (48467)

concentration

2000 μg/mL in methanol

application(s)

environmental

Analyte

Description

    Benzidine
    3,3′-Dichlorobenzidine

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service