Skip to Content
Merck
All Photos(1)

Key Documents

807163

TSKgel® DEAE-SW (10 µm) HPLC Columnss

L x I.D. 7.5 cm × 7.5 mm, DEAE (diethylaminoethyl) phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

product name

TSKgel® DEAE-SW Columns, phase DEAE (diethylaminoethyl), L × I.D. 7.5 cm × 7.5 mm, 10 μm particle size

material

stainless steel hardware

product line

TSKgel®

feature

counter ion H2PO4-

manufacturer/tradename

Tosoh 07163

parameter

0-100% organic solvent
0.05-0.1 M salt concentration (starting buffer)
0.2-0.5 M salt concentration (final buffer)
1.2 mL/min flow rate
10-45 °C temperature
200 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

7.5 cm × 7.5 mm

matrix

silica particle platform

matrix active group

DEAE (diethylaminoethyl) phase

particle size

10 μm

operating pH

2.0-7.5

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel DEAE-3SW columns are packed with 10μm porous spherical silica beads which are chemically modified with a weak anion exchanger. They are used for the separation and analysis of proteins, peptides, nucleotides, nucleosides and other biologically active molecules.

Physical form

Shipped in methanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service