Skip to Content
Merck
All Photos(1)

Key Documents

442349

Supelco

2-Fluoronaphthalene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

Fc1ccc2ccccc2c1

InChI

1S/C10H7F/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7H

InChI key

BAGQBTMEEISJLK-UHFFFAOYSA-N

General description

2-Fluoronaphthalenes are mono-substituted naphthalenes, which belong to the class of polycyclic aromatic hydrocarbons (PAHs).[1]

Application

2-Fluoronaphthalene has been used as a reference standard for the determination of the analyte in duloxetine hydrochloride active pharmaceutical ingredient (API) by reversed-phase high-performance liquid chromatography (RP-HPLC).[2] It has also been used as an internal standard for the determination of fluoride in biological samples by gas chromatography-mass spectrometry (GC-MS).[3][4]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensitive determination of fluoride in biological samples by gas chromatography-mass spectrometry after derivatization with 2-(bromomethyl) naphthalene
Kwon SM and Shin HS
Analytica Chimica Acta, 852(8), 162-167 (2014)
A Validated RP-HPLC Method for the Analysis of 1-fluoronaphthalene and its Process-Related Impurities
Karagiannidou E, et al.
Journal of Chromatographic Science, 53(8), 1296-1302 (2015)
Resonant two-photon mass-analyzed threshold ionization spectroscopy of 1-fluoronaphthalene and 2-fluoronaphthalene
Tzeng SY, et al.
Journal of Molecular Spectroscopy, 281, 40-46 (2012)
Simple determination of fluoride in biological samples by headspace solid-phase microextraction and gas chromatography-tandem mass spectrometry
Kwon SM and Shin HS
Journal of Chromatography A, 1407(8), 216-221 (2015)
Kunal Roy et al.
European journal of medicinal chemistry, 44(5), 1941-1951 (2008-12-27)
A series of naphthalene and non-naphthalene derivatives (n=42) having cytochrome P450 2A6 and 2A5 inhibitory activities, reported by Rahnasto et al., were subjected to QSAR and QAAR studies. The analyses were performed using electronic, spatial, shape and thermodynamic descriptors to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service