30240-U
Supel™-Inert Multi-Layer Foil
maximum volume 10 L, Push/Pull Lock Valve (PLV), pkg of 10 bags
Synonym(s):
Supel™-Inert Foil 10L PLV Gas Sampling Bag with Thermogreen® LB-2 Septa
About This Item
Recommended Products
Agency
NIOSH 6603
OSHA ID-172,ID-210
description
Chemically Inert with Light and Moisture Protection
septum type: Thermogreen® LB-2 septa; non-replaceable
product line
Supel™
packaging
pkg of 10 bags
technique(s)
whole air sampling: suitable
maximum volume
10 L
size × W × L
× 12 in. × 21 in.
suitability
suitable for (Biogas and Landfill Gas (LFG))
suitable for (Calibration Mixes and Gas Transfer)
suitable for (Low molecular weight compounds such as methane, CO, CO2, and permanent gases)
suitable for
application(s)
air monitoring
environmental
industrial hygiene
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Key Features and Benefits:
- Multi-layer foil composed of two outer layers of aluminum film providing a great barrier to gases permeating through walls of the bag
- Proven low-bleed Thermogreen LB-2 septa installed in the valve fitting
- Chemically inert with moisture and light protection
- Not recommended for low ppm VOCs due to background levels (we recommend the SupelInert PVDF Tedlar alternative film for VOCs)
- Two valve choices available: Screw Cap Valve (SCV) and Push/Pull Lock Valve (PLV)
- 4 sizes: 1 L, 2 L, 5 L, and 10 L
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service