29118-U
Vials, open-top screw cap, 2 mL large opening with closures, unassembled convenience packs
volume 2 mL, clear glass vial, thread for 10-425, black polypropylene cap, PTFE/silicone septa, pkg of × 100 ea
About This Item
material
PTFE/silicone septa
black polypropylene cap
clear glass vial
description
unassembled
feature
closure type black polypropylene hole cap
closure type screw top vial
packaging
pkg of × 100 ea
O.D. × H × I.D.
12 mm × 32 mm × 6 mm
volume
2 mL
fitting
thread for 10-425
Looking for similar products? Visit Product Comparison Guide
Related Categories
Suitability
PerkinElmer Autosystem
PE ISS-100, 200, Integral 4000 and Series 200
Shimadzu GC and LC Autosamplers
Spark Marathon, Midas, Triathlon
Thermo HPLC Spectra System
Waters Alliance
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service