27230-U
Aluminum crimp seal (without septum), silver aluminum
open center, diam. × opening 20 mm × 9.5 mm, pkg of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
silver aluminum
feature
open center
packaging
pkg of 100 ea
O.D.
20 mm
diam. × opening
20 mm × 9.5 mm
fitting
thread for 20 mm
compatibility
for use with 20 mm crimp seal vial
Looking for similar products? Visit Product Comparison Guide
Related Categories
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service