Skip to Content
Merck
All Photos(1)

Documents

22075-U

Swagelok® Reducing Union

Swagelok® 400-6-1, stainless steel, 1/4 in. Swagelok, 1/16 in. Swagelok

Synonym(s):

Swagelok® 400-6-1,SS REDUCING UNION 1/4X1/16 EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
eCl@ss:
32035302
NACRES:
SB.54

material

stainless steel

packaging

pkg of 1 ea

manufacturer/tradename

Swagelok® 400-6-1

fitting

1/16 in. Swagelok
1/4 in. Swagelok

Looking for similar products? Visit Product Comparison Guide

General description

Reducing unions of Swagelok® type are mostly used as metal column terminators. All original reducing unions have a bore in the centre of about 6mm in length and an internal diameter of 1.2mm. Moreover, reducing unions have a large bore at the non-reducing site. Modification is necessary in reducing union in order to minimize peak broadening, pressure resistance and to ease the installation process.
Nuts and ferrules included.

Legal Information

Swagelok is a registered trademark of Swagelok Company

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Instrumentation for High Performance Liquid Chromatography.
Huber JFK
Science, 84-88 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service