Skip to Content
Merck
All Photos(1)

Key Documents

Z767549

Cellometer® ViaStain PI (propidium iodide) staining solution in PBS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

packaging

pack of 1 ea

manufacturer/tradename

Nexcelom CS1-0109-5ML

Legal Information

Cellometer is a registered trademark of Nexcelom Bioscience, LLC
ViaStain is a trademark of Nexcelom Bioscience, LLC

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Muta. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service