Skip to Content
Merck
All Photos(1)

Key Documents

Z744244

Wilmad® Single Crystal Mounting Rod

size 4 mm, pack of 1

Synonym(s):

EPR Mounting Rod, Suprasil Mounting Rod

Sign Into View Organizational & Contract Pricing

Select a Size

13 EACH
₹67,465.80

₹67,465.80


Estimated to ship on24 June 2025Details



Select a Size

Change View
13 EACH
₹67,465.80

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

₹67,465.80


Estimated to ship on24 June 2025Details


packaging

pack of 1

manufacturer/tradename

Wilmad WG-856-Q

size

4 mm

Looking for similar products? Visit Product Comparison Guide

General description

These rods are manufactured with the highest grade of synthetic quartz to eliminate E′ center defects and paramagnetic material contaminations.

Application

EPR

Legal Information

Wilmad is a registered trademark of SP Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service