Z739472
Labnet MultiGene™ Gradient PCR Thermal Cycler
with 96 well block (12 8 x 0.2 mL strip tubes, 96 x 0.2 mL), AC/DC input 230 V AC
Synonym(s):
gradient thermal cyclers, pcr thermal cyclers, pcr thermo cyclers, thermal cyclers, thermo cyclers
About This Item
Recommended Products
feature
CE compliant
manufacturer/tradename
Labnet TC9600-G-230V
technique(s)
PCR: suitable
Looking for similar products? Visit Product Comparison Guide
General description
- Sample Capacity: 1 x 96 well plate, 12 8 x 0.2 mL strip tubes, 96 x 0.2 mL tubes, 60 x 0.5 mL tubes
- Programmable Temperature Range: 4°C - 99.9°C
- Temperature Accuracy/Uniformity: ±0.5°C/±0.5°C
- Maximum Heating/Cooling Rate: 3°C/2°C per second
- Gradient Temperature Range: 30°C to 99°C
- Maximum Gradient Temperature Difference: 24°C
- Gradient Capability: 12 rows (horizontal), 11 rows (60 well version)
- Programmable Lid Temperature: 60°C to 65°C, 100°C to 115°C
- Program Memory: 100 complete programs
- Temperature Increments/Decrements: Yes
- Time Increments/Decrements: Yes
- Temperature control: Calculated or block
- User program folders: Yes
- Password protected programs: Yes
- Communication: USB and RS232 ports
- Dimensions (W x D x H): 9.5 x 16.5 x 9.8 in.; 24 x 42 x 25 cm
- Weight: 20 lb/9 kg
PRECISION THERMAL CONTROL: Rapid heating and cooling of the precision machined sample block is achieved by six Peltier modules. Temperature sensors across the block feed data to the microprocessor to control operation of the system and ensure accuracy and uniformity. Two types of temperature control are selectable. Block control, uses the actual block temperature to adjust heating/cooling input. Tube control, sophisticated algorithm program that calculates sample temperature based on volume and triggers heating/cooling accordingly to provide quick temperature change in the sample and minimize over- and undershoot.
EASY PROGRAMMING: The large, backlit LCD displays alphanumeric characters and a graphical representation of program steps. Simple, on-screen instructions guide the user through the programming process. 99 programs may be stored in the system memory and organized in public folders or user folders, which can be password protected.
PROGRAMMING FEATURES: A variety of applications from single temperature soaks to sophisticated multistep programs. All of these are easily set up. Time or temperature of a cycling step can be automatically increased or decreased during successive cycles.Programs may also be paused while running. During operation, actual times and temperatures are displayed. Estimated run times are automatically calculated and a log of the last run can be displayed or downloaded to a PC.
GRADIENT CAPABILITY: Can be programmed to operate with uniform temperature across the block for consistent results, or with a temperature gradient for protocol optimization. The MultiGene′s gradient is spread horizontally allowing twelve temperatures (eleven for 60 well block) to be evaluated simultaneously.
SLIDING, ADJUSTABLE HEATED LID: The heated lid, a standard feature, is fully adjustable to provide the proper pressure for use with different height tubes as well as plates. A compression mat is provided to ensure even pressure when using plates. The lid can also be flipped up for cleaning.
PRECISION THERMAL CONTROL: Rapid heating and cooling of the precision machined sample block is achieved by six Peltier modules. Temperature sensors across the block feed data to the microprocessor to control operation of the system and ensure accuracy and uniformity. Two types of temperature control are selectable. The first option is block control, which uses the actual temperature of the block to adjust heating and cooling input. THe second option is tube control, which uses a sophisticated algorithm program that calculates sample temperature based on volume and triggers heating and cooling accordingly to provide quick temperature change in the sample and minimize over- and undershoot.
EASY PROGRAMMING: Programming the MultiGene Gradient thermal cycler is very intuitive. The large, backlit LCD displays alphanumeric characters and a graphical representation of program steps. Simple, on-screen instructions guide the user through the programming process. The MultiGene Gradient is provided with common pre-programmed protocols which are easily edited by navigating to the appropriate parameter with the arrow keys and entering a new value. A maximum of 99 programs may be stored in the system memory and organized in public folders or user folders, which can be password protected.
PROGRAMMING FEATURES: A variety of applications can be performed with the MultiGene Gradient thermal cycler, from single temperature soaks to sophisticated multistep programs. All of these are easily set up. Time or temperature of a cycling step can be automatically increased or decreased during successive cycles. This is useful for "touchdown" applications and extending annealing times as enzyme is depleted. Programs may also be paused while running. During operation, actual times and temperatures are displayed. Estimated run times are automatically calculated and a log of the last run can be displayed or downloaded to a PC.
GRADIENT CAPABILITY: The MultiGene Gradient thermal cycler block can be programmed to operate with uniform temperature across the block for consistent results, or with a temperature gradient for protocol optimization. The MultiGene′s gradient is spread horizontally allowing twelve temperatures(Eleven for 60 well block) to be evaluated simultaneously. To program a gradient, the high and low temperature are set, and the software automatically calculates and displays the temperature in each row. A maximum spread of 24°C can be set up across the block.
SLIDING, ADJUSTABLE HEATED LID: The heated lid, a standard feature of the MultiGene Gradient thermal cycler, is fully adjustable to provide the proper pressure for use with different height tubes as well as plates. A compression mat is provided to ensure even pressure when using plates. For optimum performance, temperature of the lid may be programmed. To ensure that users never come in contact with the hot surface, the lid slides back and away from the samples. In addition to eliminating the risk of burns, this also provides complete access to samples. The lid can also be flipped up for cleaning.
Legal Information
comparable product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service