Key Documents
Z645494
Nichiryo pipette tips
maximum volume 1000 μL, universal, sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
colorless
sterility
sterile
feature
barrier: no
packaging
pack of 1000 ea (10 racks of 100 tips)
manufacturer/tradename
Nichiryo BMT-LRS
availability
not available in (France)
maximum volume
1000 μL
Features and Benefits
  • RNase and DNase free
  • Fits most pipettes
  • Premium packaging featuring stand-up and puncture-resistant bags
  • Racked tips are offered in both hinged and lift-off tops
Other Notes
Browse all pipette tips
Choose from one of the most recent versions:
Certificates of Analysis (COA)