Z627623
BRAND® Dispensette® Organic bottle-top dispenser, analog-adjustable volume
1-10 mL, with SafetyPrime
Synonym(s):
bottle top dispensers
Select a Size
₹40,074.15
Select a Size
About This Item
₹40,074.15
Recommended Products
feature
DE-M marking
with SafetyPrime
manufacturer/tradename
BRAND 4730141 EU
BrandTech 4731141 US
measuring range
1-10 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Easy to dismantle for cleaning, with replaceable filling valves
- Autoclavable at 121°C
- Fast volume adjustment with analog slide
- Simple calibration adjustment with supplied tool
- SafetyPrime™ recirculation valve reduces reagent waste and the safety discharge system reduces the risk of inadvertent dispensing and splashes
- To allow the bottle label to face the user, the valve block can be rotated 360°
- A telescopic filling tube adjusts easily to different size bottles
The parts of the Dispensette Organic which are in contact with medium are made of: Borosilicate glass, ceramic, tantalum, ETFE, FEP & PFA. The discharge tube safety screw cap is PP.
Operating limits:
- Vapor pressure max. 500 mbar
- Viscosity max. 500 mm2/s
- Temperature max. 40°C
- Density max. 2.2 g/cm3
- Accuracy: ±0.5%, precision ±0.1%
Features and Benefits
- Simple: Wide range of applications for organic solvents, acids, alkalis and saline solutions.
- Easy: Minimum operating forces, maximum comfort.
- Efficient: Reliable in challenging continuous operation and with aggressive media.
Linkage
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service