Skip to Content
Merck
All Photos(1)

Key Documents

Z379522

Dust-Pro pressurized duster

weight 8 oz, pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41103200

packaging

pkg of 1 ea

weight

8 oz

General description

Contents are 100% non-flammable, ozone-safe (no CFC′s), and triple-filtered for purity. Gas stream is delivered through narrow-orifice nozzle for dusting with pinpoint accuracy.

Packaging

Package size: 8 oz

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service