Documents
Z370789
Sigma-Aldrich
Scienceware® cloning cylinders, polystyrene
40 of each size
Synonym(s):
cloning cylinder, cloning equipment
About This Item
UNSPSC Code:
41106618
NACRES:
NB.22
sterility
sterile
packaging
box of 3 bags
manufacturer/tradename
Bel-Art H37847-0000
General description
To isolate a clone, sterile grease (not included) is applied to bottom edge of cylinder and inverted over clone of choice. Provides an isolation chamber to trypsinize and recover clonal colonies.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP