Key Documents
Z329789
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 500 mL, label, Acetone, without venting valve
Select a Size
5 EA
₹5,126.00
₹5,126.00
Please contact Customer Service for Availability
Select a Size
Change View
5 EA
₹5,126.00
About This Item
UNSPSC Code:
41103209
NACRES:
NB.17
₹5,126.00
Please contact Customer Service for Availability
material
low-density polyethylene bottle
red closure
round bottle
translucent bottle
wide-mouth bottle
feature
label Acetone
with cap
without venting valve
packaging
pack of 5 ea
manufacturer/tradename
BRAND 144085
capacity
500 mL
General description
Flexible, high-quality wash bottles with integral wash steam and sealing cone in the closure. Large opening for convenient filling. Label with designation of contents and general safety statements (with CAS no. and NFPA codes).
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
Other Notes
BR144085 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
accessory
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)