Skip to Content
Merck
All Photos(1)

Key Documents

Z328170

BRAND® Transferpette® 12-channel, digital adjustable pipette

volume 2-20 μL, accuracy: 1.0%, precision: 0.6%

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
₹76,521.93

₹76,521.93


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
₹76,521.93

About This Item

UNSPSC Code:
41121500

₹76,521.93


Please contact Customer Service for Availability

feature

12-channel
electronic: no
recommended tips Nano-cap, 20 μL
* 0.5-10μL

subdivision 0.1 μL

manufacturer/tradename

BrandTech 703622

volume measuring range

2-20 μL, accuracy: 1.0%, precision: 0.6%

volume

20 μL

Looking for similar products? Visit Product Comparison Guide

General description

The Transferpette-12 is designed with ease in mind.The contoured handle and accessible controls work naturally in your hand. The light weight yet durable construction and low operating force minimize strain and fatigue.

Special features: FKM V-rings and a stepped tip ejector significantly reduce the ejection force, easy to maintain with replaceable individual tip cones and seals, universal tip cones - fit all convential pipette tips, corrosion-resistant piston, Easy Calibration Technique: No tools needed to make adjustments, the freely rotatble manifold can be entirely autoclaved at 121°C, separate ejector function.

* Filter tips

Packaging

Transferpette-12, conformity certified with performance certificate, 1 Tip-Box, filled with PLASTIBRAND pipette tips, 1 Tip-Rack for refilling, 1 stamd, 1 reagent-reservoir, 1 set of sealing rings made of FKM.

Other Notes

BR703622 (India)

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service