Z327891
BRAND® Transferpette® pipette, fixed volume, single channel
volume 200 μL, accuracy: 0.6%, precision: 0.2%
Select a Size
₹11,799.25
Select a Size
About This Item
₹11,799.25
description
corrosion resistant piston
supplied with performance certificate and calibration certificate.
feature
electronic: no
recommended tips 50-1000 μL & 50-1250 μL
single-channel
manufacturer/tradename
BRAND
BrandTech 704146
volume measuring range
200 μL, accuracy: 0.6%, precision: 0.2%
volume
200 μL
General description
Changing volume is easy without snagging gloves. Detents prevent inadvertent volume changes. Easy to calibration tool supplied. The slim shaft (autoclavable at 121°C) makes it possible to pipette in the narrowest vessels without removing the tip ejector. The taper is designed to fit PLASTIBRAND and most other leading manufacturers tips.
CE-marked according to IVD-Directive 98/79 CE.
* Filter tips
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)