Skip to Content
Merck
All Photos(1)

Key Documents

X1254

Sigma-Aldrich

Anti-XPA (C-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-XP1, Anti-XPAC, Anti-Xeroderma pigmentosum, complementary group A

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
₹65,256.90

₹65,256.90


Estimated to ship on19 June 2025Details


Request a Bulk Order

Select a Size

Change View
200 μL
₹65,256.90

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

₹65,256.90


Estimated to ship on19 June 2025Details


Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 35 kDa (doublet)

species reactivity

human

technique(s)

western blot: 1:1,000-1:2,000 using Jurkat cell lysates
western blot: 1:250-1:500 using Rat1 cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XPA(7507)
mouse ... Xpa(22590)
rat ... Xpa(298074)

General description

Xeroderma pigmentosum group A-complementing protein (XPA) or DNA repair protein complementing XP-A cells human excision repair protein. XPA encodes a hydrophilic metalloprotein. It displays a C4 zinc finger motif, a central globular domain and has disordered regions in the N and C-terminus. The gene XPA is localized in human chromosome 9q22.33.

Specificity

Anti-XPA (C-terminal) specifically recognizes human XPA.

Immunogen

synthetic peptide corresponding to amino acids 257-273 of human XPA, conjugated to KLH via an N-terminal added cysteine residue. The immunizing peptide differs from the mouse and rat sequences by one amino acid.

Application

Anti-XPA (C-terminal) antibody produced in rabbit has been used in immunolabeling and immunoblotting.

Biochem/physiol Actions

Xeroderma pigmentosum group A-complementing protein (XPA) interacts with nucleotide excision repair (NER) subunits, such as replication protein A (RPA), excision repair complementing 1 protein (ERCC1), and transcription factor II (TFIIH). However, it is widely accepted that the Xeroderma pigmentosum group C-complementing protein (XPC)-human Rad23 homolog (hHR23B) complex recognizes the DNA damage-induced helical distortion. After this, the transcription factor IIH (TFIIH), XPA (possibly in its homodimeric form), and replication protein A (RPA) arrive sequentially at the site of damage. XPA interacts directly with DNA via the zinc finger motif. RPA coordinates with XPA in the positioning in the nucleotide excision repair (NER) bubble. Defects in the excision repair leads to photosensitivity syndrome called xeroderma pigmentosum (XP).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chin-Ju Park et al.
The FEBS journal, 273(8), 1600-1608 (2006-04-21)
Xeroderma pigmentosum (XP) is an inherited disease in which cells from patients exhibit defects in nucleotide excision repair (NER). XP proteins A-G are crucial in the processes of DNA damage recognition and incision, and patients with XP can carry mutations
Reinhart Speeckaert et al.
Pigment cell & melanoma research, 27(4), 512-524 (2014-03-13)
Hyperpigmentation is a key feature in a variety of inherited and acquired syndromes. Nonetheless, determining the exact diagnosis only on the clinical phenotype can be challenging, and a detailed search for associated symptoms is often of crucial importance. As pigmentation
Paul Verbruggen et al.
PLoS computational biology, 10(1), e1003438-e1003438 (2014-02-07)
DNA repair and other chromatin-associated processes are carried out by enzymatic macromolecular complexes that assemble at specific sites on the chromatin fiber. How the rate of these molecular machineries is regulated by their constituent parts is poorly understood. Here we
Effects of tryptophan and portocaval anastomosis on activity and brain tryptophan metabolism [proceedings].
D L Bloxam et al.
British journal of pharmacology, 60(2), 277P-277P (1977-06-01)
Lucia Borszéková Pulzová et al.
International journal of molecular sciences, 21(6) (2020-04-03)
The nucleotide excision repair (NER) pathway is activated in response to a broad spectrum of DNA lesions, including bulky lesions induced by platinum-based chemotherapeutic agents. Expression levels of NER factors and resistance to chemotherapy has been examined with some suggestion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service