All Photos(1)
About This Item
Linear Formula:
C9H10N2O8PNa
CAS Number:
Molecular Weight:
328.15
EC Number:
UNSPSC Code:
12352204
PubChem Substance ID:
storage temp.
−20°C
InChI
1S/C9H11N2O8P.Na.H/c12-3-4-6-7(19-20(15,16)18-6)8(17-4)11-2-1-5(13)10-9(11)14;;/h1-2,4,6-8,12H,3H2,(H,15,16)(H,10,13,14);;
InChI key
OCYALKQVLHMNOH-UHFFFAOYSA-N
Caution
Do not confuse with the common uridine 2′(3′)-monophosphate mixed isomers
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Tomasz Grabarkiewicz et al.
Journal of molecular modeling, 12(2), 205-212 (2005-09-01)
Two uridine 2',3'-cyclic monophosphate (cUMP) derivatives, 5'-deoxy (DcUMP) and 5'-O-methyl (McUMP), were studied by means of quantum chemical methods. Aqueous solvent effects were estimated based on the isodensity-surface polarized-continuum model (IPCM). Gas phase calculations revealed only slight energy differences between
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service