Skip to Content
Merck
All Photos(1)

Key Documents

U3754

Sigma-Aldrich

Uridine-2-14C

aqueous ethanol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2O6
CAS Number:
Molecular Weight:
244.20
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

aqueous ethanol solution

extent of labeling

40-60 mCi per mmol

shipped in

dry ice

storage temp.

−20°C

SMILES string

OCC1OC(C(O)C1O)N2C=CC(=O)NC2=O

Looking for similar products? Visit Product Comparison Guide

Packaging

serum bottle

Physical form

Aqueous solution containing 2% ethanol

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service