Key Documents
TMO382099-1000
Nalgene® Certified Low Particulate narrow-mouth bottle
natural high-density polyethylene, volume 1000 mL, case of 24 ea
Synonym(s):
HDPE bottle, certified nalgene bottle
Select a Size
1 EACH
₹35,241.25
₹35,241.25
Estimated to ship on07 July 2025Details
Select a Size
Change View
1 EACH
₹35,241.25
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
₹35,241.25
Estimated to ship on07 July 2025Details
material
natural high-density polyethylene
packaging
case of 24 ea
manufacturer/tradename
Nalgene 382099-1000
capacity
1000 mL
volume
1000 mL
General description
Effectively store high-purity chemicals with Thermo Scientific Nalgene Certified Low Particulate Narrow-Mouth HDPE Bottles with Closure, which are extremely low in trace metal content. Controlled environment manufacturing minimizes particulate contamination.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)