Skip to Content
Merck
All Photos(1)

Key Documents

T9266

Sigma-Aldrich

3-Thiophenecarboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4OS
CAS Number:
Molecular Weight:
112.15
Beilstein:
105889
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

vapor pressure

0.31 mmHg ( 20 °C)

autoignition temp.

>392 °F

refractive index

n20/D 1.583 (lit.)

bp

194-196 °C (lit.)
86-87 °C/20 mmHg (lit.)

density

1.28 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccsc1

InChI

1S/C5H4OS/c6-3-5-1-2-7-4-5/h1-4H

InChI key

RBIGKSZIQCTIJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service