Skip to Content
Merck
All Photos(1)

Key Documents

T7790

Sigma-Aldrich

Tyrphostin 63

≥98%

Synonym(s):

4-Hydroxybenzyl malononitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2O
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥98%

potency

6500 μM IC50 (negative control)

solubility

DMSO: soluble

storage temp.

2-8°C

SMILES string

Oc1ccc(CC(C#N)C#N)cc1

InChI

1S/C10H8N2O/c11-6-9(7-12)5-8-1-3-10(13)4-2-8/h1-4,9,13H,5H2

InChI key

PDRLQWJJFLVIRC-UHFFFAOYSA-N

Biochem/physiol Actions

EGFR tyrosine kinase inhibitor.

Preparation Note

Tyrphostins are soluble in DMSO. Although the presence of water in the solution may accelerate hydrolysis, frozen solutions in DMSO should still be stable for months.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service