Documents
T6781
Sigma-Aldrich
Gel-sealing tape
roll size 1 in. × 72 yard
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
roll size
1 in. × 72 yard
Application
Used for sealing the ends of the gel tray during gel formation.