Skip to Content
Merck
All Photos(1)

Key Documents

T2182

Sigma-Aldrich

Tubulozole-C hydrochloride

≥98%

Synonym(s):

R 46846

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
Pricing and availability is not currently available.

Assay

≥98%

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W de Priester et al.
European journal of cell biology, 51(1), 23-32 (1990-02-01)
Capping of Concanavalin A (Con A) receptors can be inhibited in Dictyostelium by treatment of amoebae with the microtubular drug tubulozole. In cells that were incubated with Con A or with fluorescent Con A conjugate the capping process was completed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service