Skip to Content
Merck
All Photos(1)

Key Documents

T1284

Sigma-Aldrich

Calcitonin eel

≥97% (HPLC)

Synonym(s):

Cys-Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asp-Val-Gly-Ala-Gly-Thr-Pro-NH2 [Disulfide bridge: 1–7], Thyrocalcitonin Eel

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C146H241N43O47S2
CAS Number:
Molecular Weight:
3414.87
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
form:
solid
Assay:
≥97% (HPLC)

Assay

≥97% (HPLC)

form

solid

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(NC(=O)C1CSSCC(N)C(=O)NC(CO)C(=O)NC(CC(N)=O)C(=O)NC(CC(C)C)C(=O)NC(CO)C(=O)NC(C(C)O)C(=O)N1)C(C)C)C(=O)NCC(=O)NC(CCCCN)C(=O)NC(CC(C)C)C(=O)NC(CO)C(=O)NC(CCC(N)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CC(C)C)C(=O)NC(Cc2cnc[nH]2)C(=O)NC(CCCCN)C(=O)NC(CC(C)C)C(=O)NC(CCC(N)=O)C(=O)NC(C(C)O)C(=O)NC(Cc3ccc(O)cc3)C(=O)N4CCCC4C(=O)NC(CCCNC(N)=N)C(=O)NC(C(C)O)C(=O)NC(CC(O)=O)C(=O)NC(C(C)C)C(=O)NCC(=O)NC(C)C(=O)NCC(=O)NC(C(C)O)C(=O)N5CCCC5C(N)=O

InChI

1S/C146H241N43O47S2/c1-66(2)47-88(175-140(231)112(72(13)14)184-137(228)100-64-238-237-63-81(149)119(210)178-97(60-190)135(226)174-94(54-105(152)200)132(223)172-92(51-70(9)10)130(221)180-99(62-192)136(227)187-115(76(18)195)143(234)181-100)120(211)159-58-107(202)163-82(27-20-22-42-147)121(212)169-91(50-69(7)8)129(220)179-98(61-191)134(225)167-85(36-39-103(150)198)123(214)165-87(38-41-109(204)205)124(215)171-90(49-68(5)6)128(219)173-93(53-79-56-156-65-161-79)131(222)164-83(28-21-23-43-148)122(213)170-89(48-67(3)4)127(218)166-86(37-40-104(151)199)126(217)186-114(75(17)194)142(233)177-96(52-78-32-34-80(197)35-33-78)144(235)189-46-26-31-102(189)138(229)168-84(29-24-44-157-146(154)155)125(216)185-113(74(16)193)141(232)176-95(55-110(206)207)133(224)183-111(71(11)12)139(230)160-57-106(201)162-73(15)118(209)158-59-108(203)182-116(77(19)196)145(236)188-45-25-30-101(188)117(153)208/h32-35,56,65-77,81-102,111-116,190-197H,20-31,36-55,57-64,147-149H2,1-19H3,(H2,150,198)(H2,151,199)(H2,152,200)(H2,153,208)(H,156,161)(H,158,209)(H,159,211)(H,160,230)(H,162,201)(H,163,202)(H,164,222)(H,165,214)(H,166,218)(H,167,225)(H,168,229)(H,169,212)(H,170,213)(H,171,215)(H,172,223)(H,173,219)(H,174,226)(H,175,231)(H,176,232)(H,177,233)(H,178,210)(H,179,220)(H,180,221)(H,181,234)(H,182,203)(H,183,224)(H,184,228)(H,185,216)(H,186,217)(H,187,227)(H,204,205)(H,206,207)(H4,154,155,157)

InChI key

JDJALSWDQPEHEJ-UHFFFAOYSA-N

Gene Information

mouse ... Edn1(13614)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Cys-Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asp-Val-Gly-Ala-Gly-Thr-Pro-NH2 [Disulfide bridge: 1–7]

Biochem/physiol Actions

Hypocalcemic hormone produced by the parafollicular C cells of the thyroid or by the ultimobranchial bodies of nonmammalian vertebrates. Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes.

Other Notes

A 32 amino acid polypeptide, 8 of which are conserved across all species.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C L H Huang et al.
Cellular and molecular biology (Noisy-le-Grand, France), 52(3), 33-43 (2007-05-31)
Calcitonin is a thirty-two amino acid peptide that contains an N-terminal disulphide bridge and a C-terminal prolineamide residue. It is released from thyroid parafollicular C-cells and its direct actions on the osteoclast account for its physiological effects whether as a
S Tanaka et al.
Cellular and molecular biology (Noisy-le-Grand, France), 52(3), 19-23 (2007-05-31)
Calcitonin is a calcium-regulating peptide hormone that acts directly on osteoclasts and suppresses their function. In this review we will focus on the downstream signaling pathways of calcitonin receptor regulating cytoskeletal organization of osteoclasts. We will also introduce the clinical
F J A de Paula et al.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme, 42(5), 299-306 (2010-03-11)
This review reflects on the past, present, and future of translational research on calcitropic hormones and bone metabolism. Calcitonin (CT) and parathormone (PTH) are complementary hormones involved in the acquisition and maintenance of bone mass and regulation of calcium metabolism.
D Naot et al.
Bone, 43(5), 813-818 (2008-08-09)
The 'calcitonin family' is a group of peptide hormones that share structural similarities with calcitonin, and includes calcitonin gene-related peptide (CGRP), amylin, adrenomedullin and adrenomedullin 2 (intermedin). These hormones are produced by different tissues, with calcitonin being produced in thyroid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service