Skip to Content
Merck
All Photos(1)

Documents

SRP8027

Sigma-Aldrich

CD24 (mouse): FC (human)

recombinant, expressed in HEK 293 cells, >90% (SDS-PAGE)

Synonym(s):

M1/69-J11D heat stable antigen, NaMN adenylyltransferase 1, Nectadrin, X62 Heat stable antigen (HSA)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in HEK 293 cells

Assay

>90% (SDS-PAGE)

form

lyophilized

mol wt

~50 kDa by SDS-PAGE

packaging

pkg of 50 μg

impurities

<0.06 EU/μg endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

CD24 is a highly glycosylated protein expressed as glycosyl-phosphatidyl-inositol (GPI) anchored molecule. Widely expressed, mainly on T cells, B cells and is upregulated in numerous types of cancers. It has costimulatory activity for antigen specific T cell responses. Polymorphisms of human CD24 are associated with several autoimmune diseases. Recently a new role as negative regulator of the immune response has been reported. CD24-deficient mice are more sensitive to inflammatory responses. CD24 together with the membrane protein SIGLEC interacts with danger-associated molecular patterns (DAMPs), such as HMGB1, HSP90 or HSP70. CD24/SIGLEC complex may differentially regulate the host response to DAMPs and PAMPs (pathogen-associated molecular patterns).

Physical form

Lyophilized from PBS.

Reconstitution

Reconstitute with 50 μL deionized water.

Other Notes

Mouse CD24 (aa 27-52) is fused at the C-terminus to the Fc portion of human IgG1.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service