FGF14 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. A mutation in this gene is associated with autosomal dominant cerebral ataxia. Alternatively spliced transcript variants have been found for this gene. Recombinant human FGF14 protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.
Physical form
0.25 mg/mL in 20 mM Tris-HCl buffer (pH 8.0) containing 0.2 M NaCl, 5 mM DTT and 50% glycerol.