Skip to Content
Merck
All Photos(2)

Key Documents

SRP5015

Sigma-Aldrich

CK1 ε, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CK1E, CSNK1E, HCKIE, MGC10398

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

39-53 nmol/min·mg

mol wt

~72 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CSNK1E(1454)

General description

CK1ε is a member of the CK1 family of serine/threonine protein kinases which play an important role in diverse cell processes, including DNA replication and repair. CK1ε is a regulator of Yes-associated protein (YAP) transcription coactivator which is a key regulator of organ size and a candidate human oncogene. CK1ε is activated by CCK2R and this then phosphorylates PKD2 at Ser244. Phosphorylation of PKD2 leads to its nuclear accumulation and efficient phosphorylation of nuclear PKD2 substrates in human gastric cancer cells. CKIε can phosphorylate topoisomerase (topo) IIalpha at serine-1106 and this regulates the enzyme activity and sensitivity to topo II-targeted drugs.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian G Grozav et al.
Nucleic acids research, 37(2), 382-392 (2008-12-02)
We previously reported that phosphorylation of topoisomerase (topo) IIalpha at serine-1106 (Ser-1106) regulates enzyme activity and sensitivity to topo II-targeted drugs. In this study we demonstrate that phosphorylation of Ser-1106, which is flanked by acidic amino acids, is regulated in
J von Blume et al.
The EMBO journal, 26(22), 4619-4633 (2007-10-27)
Protein kinase D2 (PKD2), a member of the PKD family of serine/threonine kinases, is localized in various subcellular compartments including the nucleus where the kinase accumulates upon activation of G-protein-coupled receptors. We define three critical post-translational modifications required for nuclear

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service