Skip to Content
Merck
All Photos(1)

Documents

SRP0675

Sigma-Aldrich

IL-17A

recombinant, expressed in E. coli

Synonym(s):

CTLA8, Cytotoxic T-Lymphocyte-Associated Antigen 8, IL-17, IL17, Interleukin-17A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in E. coli

form

frozen liquid

mol wt

dimer 35 kDa
monomer 17.5 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IL17A(3605)

General description

Fc region of human immunoglobulin G1 (IgG1) protein, also known as Il-4, GenBank Accession No. P01857, a.a. 100-330, expressed in an HEK293 cell expression system. MW = 27 kDa.
Human secreted interleukin 17 A (IL-17A) also known as, IL-17 and cytotoxic T lymphocyte antigen 8 (CTLA8) GenBank Accession No. NM_ 002190, a.a 20-155(end), expressed in an e. coli expression system. This protein exists naturally as a homodimer; M.W. = 17.5 kDa (monomer); 35 kDa dimer

Physical form

Aqueous solution

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaiyan Yang et al.
Journal of receptor and signal transduction research, 41(3), 284-293 (2020-08-26)
Elucidating the molecular mechanism of the migration and invasion is critical for identifying novel therapeutic targets and may significantly improve the prognosis of colorectal cancer. Emerging evidence suggests an involvement of dysregulated microRNAs in the process of tumorigenesis. Here, we
David E Gordon et al.
Science (New York, N.Y.), 370(6521) (2020-10-17)
The COVID-19 pandemic, caused by severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), is a grave threat to public health and the global economy. SARS-CoV-2 is closely related to the more lethal but less transmissible coronaviruses SARS-CoV-1 and Middle East respiratory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service