Skip to Content
Merck
All Photos(1)

Documents

SRP0306

Sigma-Aldrich

HDAC Substrate 2, fluorogenic

≥95% (HPLC)

Synonym(s):

HDAC, histone deacetylase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥95% (HPLC)

form

aqueous solution

packaging

pkg of 250 nmol

shipped in

dry ice

storage temp.

−70°C

General description

A fluorogenic, acetylated peptide substrate for HDAC′s. It is a suitable substrate for many HDAC′s tested so far, except class 2a HDAC.

Application

Study enzyme kinetics, and screen small molecule inhibitors for drug discovery and HTS applications.

Physical form

Supplied as a 5 mM solution in DMSO.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service