Skip to Content
Merck
All Photos(1)

Key Documents

SML2862

Sigma-Aldrich

MC1742

≥98% (HPLC)

Synonym(s):

5-(4-(Biphenyl-4-yl)-6-oxo-1,6-dihydropyrimidin-2-ylthio)-N-hydroxypentanamide, 5-[(4-[1,1′-Biphenyl]-4-yl-1,6-dihydro-6-oxo-2-pyrimidinyl)thio]-N-hydroxypentanamide, MC 1742, MC-1742

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
₹10,738.40
25 MG
₹43,202.58

₹10,738.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
₹10,738.40
25 MG
₹43,202.58

About This Item

Empirical Formula (Hill Notation):
C21H21N3O3S
CAS Number:
Molecular Weight:
395.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

₹10,738.40


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

S(CCCCC(=O)NO)c1[nH][c](cc(n1)c2ccc(cc2)c3ccccc3)=O

InChI

1S/C21H21N3O3S/c25-19(24-27)8-4-5-13-28-21-22-18(14-20(26)23-21)17-11-9-16(10-12-17)15-6-2-1-3-7-15/h1-3,6-7,9-12,14,27H,4-5,8,13H2,(H,24,25)(H,22,23,26)

InChI key

AOFVDNFTELWRHV-UHFFFAOYSA-N

Biochem/physiol Actions

Histone deacetylases inhibitor against class I (HDAC1/2/3/8), IIB (HDAC6/10), IV (HDAC11) HDACs with anti-cancer and HIV latency reativation potency.
MC1742 is a uracil-based hydroxyamide (UBHA) that acts as a subtype-selective histone deacetylases (HDACs) inhibitor (IC50: class I HDAC1/2/3/8 = 100/110/20/610 nM, class IIB HDAC6/10 = 7/40 nM, class IV HDAC11 = 100 nM, class IIA HDAC4/5/7/9 >50 μM). MC1742 exerts greater antiproliferation potency than SAHA in sarcoma cancer stem cell (CSC) cultures (MC1742/SAHA IC50 in μM = 0.25/1/RD, 1.12/1.13//MG-63, 1.4/3.92/SK-ES-1 post 72-hr treatment) and effectively reactivates HIV from latency (EC50 = 350 nM; JLAT 10.6 cells) by upregulating histone acetylation at the HIV promoter without activating T cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gemma Di Pompo et al.
Journal of medicinal chemistry, 58(9), 4073-4079 (2015-04-24)
Musculoskeletal sarcomas are aggressive malignancies of bone and soft tissues often affecting children and adolescents. Histone deacetylase inhibitors (HDACi) have been proposed to counteract cancer stem cells (CSCs) in solid neoplasms. When tested in human osteosarcoma, rhabdomyosarcoma, and Ewing's sarcoma
Elleard Fw Heffern et al.
Journal of virus eradication, 5(2), 84-91 (2019-06-14)
Current antiretroviral therapy can suppress HIV replication, increase CD4 count and result in increased lifespan. However, it cannot eradicate the virus due to the presence of latent provirus in cellular reservoirs, such as resting CD4+ T cells. Using combination latency-reversing
Tokuhiro Chano et al.
American journal of cancer research, 6(4), 859-875 (2016-05-18)
The glycolytic-based metabolism of cancers promotes an acidic microenvironment that is responsible for increased aggressiveness. However, the effects of acidosis on tumour metabolism have been almost unexplored. By using capillary electrophoresis with time-of-flight mass spectrometry, we observed a significant metabolic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service