Skip to Content
Merck
All Photos(1)

Key Documents

SML2720

Sigma-Aldrich

Miriplatin

>97.5% (Pt analysis)

Synonym(s):

Cyclohexane-(1R,2R)-diamineplatinum(II) dimyristate, MPT, SM-11355

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
₹7,936.50
50 MG
₹32,067.90

₹7,936.50


Estimated to ship on06 June 2025Details


Request a Bulk Order

Select a Size

Change View
10 MG
₹7,936.50
50 MG
₹32,067.90

About This Item

Empirical Formula (Hill Notation):
C34H68N2O4Pt
CAS Number:
Molecular Weight:
763.99
UNSPSC Code:
12352200
NACRES:
NA.77

₹7,936.50


Estimated to ship on06 June 2025Details


Request a Bulk Order

Quality Level

Assay

>97.5% (Pt analysis)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCC(O[Pt]1(N[C@@H]2CCCC[C@H]2N1)OC(CCCCCCCCCCCCC)=O)=O

InChI

1S/2C14H28O2.C6H12N2.Pt/c2*1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16;7-5-3-1-2-4-6(5)8;/h2*2-13H2,1H3,(H,15,16);5-8H,1-4H2;/q;;-2;+4/p-2/t;;5-,6-;/m..1./s1

InChI key

DTRJSYSQELJDAW-BLUNCNMSSA-L

Biochem/physiol Actions

Miriplatin (SM-11355) is a platinum (II) complex-based anti-neoplastic agent with increased lipophilicity than its family members cisplatin and carboplatin.
Platinum (II) complex-based anti-neoplastic agent with increased lipophilicity than cisplatin and carboplatin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenji Ikeda et al.
Hepatology research : the official journal of the Japan Society of Hepatology, 47(7), 632-640 (2016-08-04)
The effect of transcatheter arterial chemoembolization (TACE) is not necessarily sufficient in patients with multiple hepatocellular carcinoma (HCC). We evaluated the antitumor activity and adverse events of TACE using warmed miriplatin suspension for multiple HCC. Seventy patients with multiple HCC
Toru Ishikawa et al.
Molecular and clinical oncology, 5(5), 511-516 (2016-11-25)
Transcatheter arterial chemoembolization (TACE) is the standard therapy for Barcelona Clinic Liver Cancer (BCLC) classification intermediate stage B hepatocellular carcinoma (HCC). However, other transcatheter methods, such as transcatheter arterial infusion (TAI), also play an important role in the treatment of
Shan Liu et al.
Journal of pharmaceutical sciences, 105(1), 78-87 (2016-02-09)
Because of the insolubility of miriplatin in water, miriplatin and lipiodol suspension is the sole formulation of miriplatin approved in Japan to treat hepatocellular carcinoma by transcatheter arterial chemoembolization. Until now, there have been no reports of other pharmaceutical formulations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service