Skip to Content
Merck
All Photos(1)

Key Documents

SML2440

Sigma-Aldrich

SGC-iMLLT

≥98% (HPLC)

Synonym(s):

(S)-1-Methyl-N-(2-((2-methylpyrrolidin-1-yl)methyl)-1H-benzo[d]imidazol-5-yl)-1H-indazole-5-carboxamide, Chemical Probe 92

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
₹8,746.60
25 MG
₹35,365.28

₹8,746.60


Estimated to ship on25 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
₹8,746.60
25 MG
₹35,365.28

About This Item

Empirical Formula (Hill Notation):
C22H24N6O
CAS Number:
Molecular Weight:
388.47
UNSPSC Code:
12352200
NACRES:
NA.77

₹8,746.60


Estimated to ship on25 April 2025Details


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL

storage temp.

2-8°C

SMILES string

O=C(C1=CC2=C(N(C)N=C2)C=C1)NC3=CC=C4NC(CN5[C@@H](C)CCC5)=NC4=C3

InChI

1S/C22H24N6O/c1-14-4-3-9-28(14)13-21-25-18-7-6-17(11-19(18)26-21)24-22(29)15-5-8-20-16(10-15)12-23-27(20)2/h5-8,10-12,14H,3-4,9,13H2,1-2H3,(H,24,29)(H,25,26)/t14-/m0/s1

InChI key

QGNDVASWIHEXCL-AWEZNQCLSA-N

Biochem/physiol Actions

SGC-iMLLT (Chemical Probe 92) is a cell penetrant, potent and selective inhibitor of YEATS domain MLLT1/3 histone interaction. SGC-iMLLT downregulates MYC and DDN genes, while increases CD86 in MV4;11 AML cells.
cell penetrant, potent and selective inhibitor of YEATS domain MLLT1/3 histone interaction

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moses Moustakim et al.
Angewandte Chemie (International ed. in English), 57(50), 16302-16307 (2018-10-06)
YEATS domain (YD) containing proteins are an emerging class of epigenetic targets in drug discovery. Dysregulation of these modified lysine-binding proteins has been linked to the onset and progression of cancers. We herein report the discovery and characterisation of the
Joshua N Asiaban et al.
ACS chemical biology, 15(4), 895-903 (2020-03-17)
ENL is a transcriptional coactivator that recruits elongation machinery to active cis-regulatory elements upon binding of its YEATS domain-a chromatin reader module-to acylated lysine side chains. Discovery chemistry for the ENL YEATS domain is highly motivated by its significance in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service