Skip to Content
Merck
All Photos(1)

Documents

SML1326

Sigma-Aldrich

PK-THPP

≥98% (HPLC)

Synonym(s):

1-[1-[6-([1,1′-Biphenyl]-4-ylcarbonyl)-5,6,7,8-tetrahydropyrido[4,3-d]pyrimidin-4-yl]-4-piperidinyl]-1-butanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H32N4O2
CAS Number:
Molecular Weight:
468.59
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL, clear
DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(CCC)C(CC1)CCN1C2=C3C(CCN(C(C4=CC=C(C5=CC=CC=C5)C=C4)=O)C3)=NC=N2

InChI

1S/C29H32N4O2/c1-2-6-27(34)23-13-16-32(17-14-23)28-25-19-33(18-15-26(25)30-20-31-28)29(35)24-11-9-22(10-12-24)21-7-4-3-5-8-21/h3-5,7-12,20,23H,2,6,13-19H2,1H3

InChI key

CJZGRIRZVHNUSM-UHFFFAOYSA-N

Biochem/physiol Actions

PK-THPP is a potent antagonist of tandem pore potassium channel subunit TASK-3 (KCNK9, K2P9.1) with an IC50 value of 35 nM for TASK-3 compared to 303 nM for TASK-1 and with >140 fold selectivity over a wider range of potassium channels. PK-THPP was found to act as a respiratory stimulant, stimulating breathing in isoflurane-anesthetized rats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service