Skip to Content
Merck
All Photos(1)

Key Documents

SML0861

Sigma-Aldrich

A-484954

≥98% (HPLC)

Synonym(s):

7-Amino-1-cyclopropyl-3-ethyl-1,2,3,4-tetrahydro-2,4-dioxo-pyrido[2,3-d]pyrimidine-6-carboxamideddd, A484954

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15N5O3
CAS Number:
Molecular Weight:
289.29
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 3 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C13H15N5O3/c1-2-17-12(20)8-5-7(10(15)19)9(14)16-11(8)18(13(17)21)6-3-4-6/h5-6H,2-4H2,1H3,(H2,14,16)(H2,15,19)

InChI key

HWODCHXORCTEGU-UHFFFAOYSA-N

Application

A-484954 has been used a selective and potent inhibitor to inhibit eEF2K (eukaryotic elongation factor 2 kinase), in order to check the enhancement of synaptic transmission in the stratum radiatum of the hippocampal CA1 region. It has also been used as an eEF2K inhibitor to monitor real-time luminescence of U2OS Arntl::dLUC cells.

Biochem/physiol Actions

A-484954 is a potent, selective inhibitor of eukaryotic longation factor-2 kinase (eEF2K).
A-484954 is a potent, selective inhibitor of eukaryotic longation factor-2 kinase (eEF2K). Eukaryotic elongation factor-2, which is required for protein synthesis, is inactivated upon phosphorylation by eEF2K.

Features and Benefits

This compound is featured on the Ca/CaMKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acid Suspends the Circadian Clock in Hypoxia through Inhibition of mTOR
Walton Z E, et al.
Cell (2018)
Potentiation of Schaffer-Collateral CA1 Synaptic Transmission by eEF2K and p38 MAPK Mediated Mechanisms
Weng W, et al.
Frontiers in Cellular Neuroscience, 10, 247-247 (2016)
Kanzo Suzuki et al.
Cell reports, 37(5), 109918-109918 (2021-11-04)
Ketamine is a noncompetitive glutamatergic N-methyl-d-aspartate receptor (NMDAR) antagonist that exerts rapid antidepressant effects. Preclinical studies identify eukaryotic elongation factor 2 kinase (eEF2K) signaling as essential for the rapid antidepressant action of ketamine. Here, we combine genetic, electrophysiological, and pharmacological
Zandra E Walton et al.
Cell, 174(1), 72-87 (2018-06-05)
Recent reports indicate that hypoxia influences the circadian clock through the transcriptional activities of hypoxia-inducible factors (HIFs) at clock genes. Unexpectedly, we uncover a profound disruption of the circadian clock and diurnal transcriptome when hypoxic cells are permitted to acidify

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service