Skip to Content
Merck
All Photos(1)

Key Documents

SML0022

Sigma-Aldrich

IMB-10

≥98% (HPLC)

Synonym(s):

3-(2-methylphenyl)-5-(3-phenyl-2-propen-1-ylidene)-2-thioxo-4-thiazolidinone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
₹9,634.25

₹9,634.25


Estimated to ship on30 April 2025Details

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
₹9,634.25

About This Item

Empirical Formula (Hill Notation):
C19H15NOS2
CAS Number:
Molecular Weight:
337.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

₹9,634.25


Estimated to ship on30 April 2025Details

New, lower price on this item!

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: ≥25 mg/mL

storage temp.

2-8°C

SMILES string

Cc1ccccc1N2C(=S)S\C(=C\C=C\c3ccccc3)C2=O

InChI

1S/C19H15NOS2/c1-14-8-5-6-12-16(14)20-18(21)17(23-19(20)22)13-7-11-15-9-3-2-4-10-15/h2-13H,1H3/b11-7+,17-13+

InChI key

UYZLENPCNZVWQZ-QHMSGILQSA-N

Application

IMB-10 may be used to study integrin-mediated signaling in tumor cells.

Biochem/physiol Actions

IMB-10 is a ligand for the αMβ2 integrin that stabilizes the binding of the integrin to endogenous ligands such as proMMP-9 and fibrinogen. IMB-10 blocks migration of lymphocyte cell lines and inhibits leukocyte infiltration in a murine thioglycolate-induced peritonitis.
IMB-10 is a thioxothiazolidine compound that inhibits the infiltration of leukocytes to the site of tumor and blocks the invasion of HSC-3 carcinoma. It modulates the host inflammatory responses and tumor progression.[1]
Stabilizing Ligand of αMβ2 integrin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juho Suojanen et al.
Anticancer research, 27(6B), 3775-3781 (2008-01-30)
Leukocyte immunomodulation has great clinical potential as a therapy of inflammatory conditions and cancer. We have recently developed leukocyte alphaMbeta2 integrin targeting small molecule (IMB-10) capable of inhibiting leukocyte migration and recruitment in vitro and in vivo. The purpose of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service