Skip to Content
Merck
All Photos(1)

Documents

SMB00258

Sigma-Aldrich

Eleuthoside B

≥95% (LC/MS-ELSD)

Synonym(s):

Eleutherol-4-O-β-D-gentiobioside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H32O14
CAS Number:
Molecular Weight:
568.52
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1cccc2cc3C(=O)O[C@H](C)c3c(O[C@@H]4O[C@H](CO[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)c12

InChI

1S/C26H32O14/c1-9-15-11(24(34)37-9)6-10-4-3-5-12(35-2)16(10)23(15)40-26-22(33)20(31)18(29)14(39-26)8-36-25-21(32)19(30)17(28)13(7-27)38-25/h3-6,9,13-14,17-22,25-33H,7-8H2,1-2H3/t9-,13-,14-,17-,18-,19+,20+,21-,22-,25-,26+/m1/s1

InChI key

PAEOIXITAYMQRF-ZAEQARNUSA-N

General description

Eleuthoside B is a cytotoxic marine natural product. It is an aromatic diterpenoid glycoside isolated from the soft coral Eleutherobia aurea and bulbs of Eleutherine palmifolia (Iridaceae).
Natural product derived from plant source.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indonesian medicinal plants. XX. Chemical structures of eleuthosides A, B, and C, three new aromatic glucosides from the bulbs of Eleutherine palmifolia (Iridaceae).
Hirotaka S, et al.
Chemical & Pharmaceutical Bulletin, 45(7), 1130-1134 (1997)
Sarcodictyin A and two novel diterpenoid glycosides, eleuthosides A and B, from the soft coral Eleutherobia aurea.
Sharon K, et al.
Journal of Natural Products, 59(9), 873-875 (1996)
Total synthesis of eleutherobin and eleuthosides A and B.
Nicolaou K C, et al.
Journal of the American Chemical Society, 120(34), 8674-8680 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service