Skip to Content
Merck
All Photos(3)

Key Documents

SAB5703042

Sigma-Aldrich

Anti-CD31/PECAM1 Antibody, clone 9K8O7, Rabbit Monoclonal

Synonym(s):

CD31, PECA1, GPIIA′, PECAM-1, endoCAM, CD31/EndoCAM

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
₹40,548.30

₹40,548.30


Estimated to ship on02 May 2025Details



Select a Size

Change View
100 μL
₹40,548.30

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

₹40,548.30


Estimated to ship on02 May 2025Details


biological source

rabbit

Quality Level

material

colorless

clone

9K8O7, monoclonal

form

liquid

mol wt

140 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
immunoprecipitation (IP): 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

AKQMPVEMSRPAVPLLNSNNEKMSDPNMEANSHYGHNDDVRNHAMKPINDNKEPLNSDVQYTEVQVSSAESHKDLGKKDTETVYSEVRKAVPDAVESRYSRTEGSLDGT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PECAM1(5175)

General description

The protein encoded by this gene is found on the surface of platelets, monocytes, neutrophils, and some types of T-cells, and makes up a large portion of endothelial cell intercellular junctions. The encoded protein is a member of the immunoglobulin superfamily and is likely involved in leukocyte migration, angiogenesis, and integrin activation.

Immunogen

PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 630-738 of CD31/PECAM1 (NP_000433.4).

Application

WB, IF/ICC, IP

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service