Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702466

Sigma-Aldrich

Anti-Prolactin Receptor Antibody, clone 9B10J3, Rabbit Monoclonal

Synonym(s):

HPRL, MFAB, hPRLrI

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
₹40,548.30

₹40,548.30


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
₹40,548.30

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

₹40,548.30


Estimated to ship on17 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

material

colorless

clone

9B10J3, monoclonal

form

liquid

mol wt

90 kDa

species reactivity

human, rat, rat

concentration

0.55 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MKENVASATVFTLLLFLNTCLLNGQLPPGKPEIFKCRSPNKETFTCWWRPGTDGGLPTNYSLTYHREGETLMHECPDYITGGPNSCHFGKQYTSMWRTYI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRLR(5618)

General description

This gene encodes a receptor for the anterior pituitary hormone, prolactin, and belongs to the type I cytokine receptor family. Prolactin-dependent signaling occurs as the result of ligand-induced dimerization of the prolactin receptor. Several alternatively spliced transcript variants encoding different membrane-bound and soluble isoforms have been described for this gene, which may function to modulate the endocrine and autocrine effects of prolactin in normal tissue and cancer. [provided by RefSeq, Feb 2011]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Prolactin Receptor (P16471).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service